Learn More
Aviva Systems Biology Corp PRR16 Antibody - middle region (ARP56956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRR16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRR16. Alternate names: DSC54, MGC104614ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62042