Learn More
Aviva Systems Biology Corp PRR5 Antibody - N-terminal region (ARP56749_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56749_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRR5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PRR5. Alternate names: FLJ20185, FLJ20185k, PP610, PROTOR1, PROTOR-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-207