Learn More
Aviva Systems Biology Corp PRR7 Antibody - middle region (ARP34723_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34723_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRR7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PRR7. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55534