Learn More
Aviva Systems Biology Corp PRRG2 Antibody - middle region (ARP44374_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44374_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRRG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRRG2. Alternate names: PRGP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-285-141