Learn More
Aviva Systems Biology Corp PRRG3 Antibody - N-terminal region (ARP49803_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49803_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRRG3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRRG3. Alternate names: MGC149510, MGC156177, TMG3, PRGP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60189