Learn More
Aviva Systems Biology Corp Prrg4 Antibody - N-terminal region (ARP49801_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prrg4. It was validated on Western Blot. . Alternate names: 9930111I18Rik, MGC117538, TMG4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-451