Learn More
Medchemexpress LLC PRRSV/CD163-IN-1 | 560995-89-3 | 98.28% | 557.62 | 25 MG
SDP
Catalog No. 5000363340
Encompass_Preferred
PRRSV/CD163-IN-1 is a PRRSV/CD163 inhibitor that can inhibit the interaction between the PRRSV glycoprotein (GP2a or GP4) and the CD163-SRCR5 domain. It can be used for the research of porcine reproductive and respiratory syndrome (PRRS).
  • Inhibits protein-protein interaction between PRRSV GP4 glycoprotein and CD163-SRCR5 domain
  • Well-tolerated by porcine alveolar macrophages (PAMs) at concentrations below 25 μM
  • Inhibits PRRSV infection of PAMs in a dose-dependent manner
  • Significantly inhibits infection caused by both type I and type II PRRSV strains
Catalog No. 50-003-63340 Supplier Medchemexpress LLC Supplier No. 25MGHY147089
PRRSV/CD163-IN-1 is a PRRSV/CD163 inhibitor that can inhibit the interaction between the PRRSV glycoprotein (GP2a or GP4) and the CD163-SRCR5 domain. It can be used for the research of porcine reproductive and respiratory syndrome (PRRS).
  • Inhibits protein-protein interaction between PRRSV GP4 glycoprotein and CD163-SRCR5 domain
  • Well-tolerated by porcine alveolar macrophages (PAMs) at concentrations below 25 μM
  • Inhibits PRRSV infection of PAMs in a dose-dependent manner
  • Significantly inhibits infection caused by both type I and type II PRRSV strains