Learn More
Medchemexpress LLC PRRVRLK acetate | 98.2% | 984.2 | 100 MG
SDP
Catalog No. 5000396976
Encompass_Preferred
PRRVRLK acetate is a peptide linker. It can be used in the creation of fusion proteins.
  • Peptide linker
  • Can be used to make fusion protein
Catalog No. 50-003-96976 Supplier Medchemexpress LLC Supplier No. HYP3438A100MG
PRRVRLK acetate is a peptide linker. It can be used in the creation of fusion proteins.
  • Peptide linker
  • Can be used to make fusion protein