Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PRRVRLK (acetate) | 98.2% | 984.20 | 5 MG
SDP

Catalog No. 5000399051
Encompass_Preferred

PRRVRLK acetate is a peptide linker that can be used to make fusion proteins.

  • Peptide linker
  • Can be used to make fusion protein
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in H₂O at 100 mg/mL
  • Soluble in PBS at 50 mg/mL

Catalog No. 50-003-99051 Supplier Medchemexpress LLC Supplier No. HYP3438A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRRVRLK acetate is a peptide linker that can be used to make fusion proteins.

  • Peptide linker
  • Can be used to make fusion protein
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in H₂O at 100 mg/mL
  • Soluble in PBS at 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.