Learn More
Aviva Systems Biology Corp PRRX2 Antibody - middle region (ARP57884_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57884_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRRX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PRRX2. Alternate names: MGC19843, PMX2, PRX2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-408