Learn More
Aviva Systems Biology Corp Prrx2 Antibody - middle region (ARP37871_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prrx2. It was validated on Western Blot. Prrx2,paired related homeobox 2,S8; Prx2; Prrx2,paired mesoderm homeobox protein 2,paired mesoderm homeobox protein 2,PRX-2,OTTMUSP00000012690,homeobox protein S8,homeo box of paired rule,surface antigen, homeo box of paired ruleThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Prx2, S8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-696