Learn More
Aviva Systems Biology Corp Prrxl1 Antibody - C-terminal region (ARP37846_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37846_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prrxl1. It was validated on Western Blot. Prrxl1,paired related homeobox protein-like 1,Drgx; Drg11; Prrxl1,dorsal root ganglia homeobox protein,dorsal root ganglia homeobox protein,homeobox protein DRG11,dorsal root ganglion 11,paired homeodomain protein DRG11,paired-related homeobox protein-like 1,paired related homeobox protein-like 1bThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Drg11, DrgxApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-690