Learn More
ATCC pRS313 phagemid; in E. coli HB101
Catalog No. 502379579
Encompass
One of a series of pBluescript-based centromere vectors (ATCC#77142-77145, 77157-77158) differing in the yeast selectable marker gene.  A YC-type centromere vector permitting visual detection of recombinants and production of ssDNA in E. coli.  It contains promoters for in vitro RNA synthesis, priming sites useful for sequencing, and encodes the lacZ alpha (lacZ?) peptide. pRSS56, constructed by ligating a PvuI fragment (bp 498-2412) of pBluescript KS+ to a PvuI fragment (bp 2850-730) of pBS(+), contains the KS MCS from pBluescript KS+ and the unique NdeI and AatII sites between bla and f1 origin of pBS(+).  A fragment (1.184 kb) containing the HIS3 gene was inserted into the NdeI site and a cassette contiaing CEN6 and the ARS associated with histone 4 (ARSH4) was inserted into the AatII site of pRSS56.  All ends were blunted.
Catalog No. 50-237-9579 Supplier ATCC Supplier No. 77142
One of a series of pBluescript-based centromere vectors (ATCC#77142-77145, 77157-77158) differing in the yeast selectable marker gene.  A YC-type centromere vector permitting visual detection of recombinants and production of ssDNA in E. coli.  It contains promoters for in vitro RNA synthesis, priming sites useful for sequencing, and encodes the lacZ alpha (lacZ?) peptide. pRSS56, constructed by ligating a PvuI fragment (bp 498-2412) of pBluescript KS+ to a PvuI fragment (bp 2850-730) of pBS(+), contains the KS MCS from pBluescript KS+ and the unique NdeI and AatII sites between bla and f1 origin of pBS(+).  A fragment (1.184 kb) containing the HIS3 gene was inserted into the NdeI site and a cassette contiaing CEN6 and the ARS associated with histone 4 (ARSH4) was inserted into the AatII site of pRSS56.  All ends were blunted.