Learn More
ATCC pRS415 phagemid; in E. coli
Catalog No. 502381480
Encompass
A YC-type shuttle vector with LEU2 selection in Saccharomyces cerevisiae, a beta-galactosidase reporter system for visual detection of recombinants, promoters for transcription in vitro, and an extensive polylinker flanked by BssHI sites.
Catalog No. 50-238-1480 Supplier ATCC Supplier No. 87520
Add to Cart
Add to Cart
A YC-type shuttle vector with LEU2 selection in Saccharomyces cerevisiae, a beta-galactosidase reporter system for visual detection of recombinants, promoters for transcription in vitro, and an extensive polylinker flanked by BssHI sites.