Learn More
ATCC pRS423 phagemid; in E. coli
Supplier:  ATCC 77104
Encompass
A YE-type (episomal) E. coli/S. cerevisiae shuttle vector permitting RNA synthesis in vitro, visual detection of recombinants, production of single stranded DNA and containing primer sites useful for sequencing
Catalog No. 50-237-9900