Learn More
Aviva Systems Biology Corp PRSS1 Antibody - C-terminal region (ARP60970_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60970_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRSS1. It was validated on Western Blot. . Alternate names: MGC120175, MGC149362, TRP1, TRY1, TRY4, TRYP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-217