Learn More
Aviva Systems Biology Corp PRSS21 Antibody - N-terminal region (ARP53665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRSS21. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PRSS21. Alternate names: ESP-1, ESP1, TEST1, TESTISINApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-868