Learn More
Aviva Systems Biology Corp PRSS22 Antibody - N-terminal region (ARP57603_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57603_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRSS22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRSS22. Alternate names: BSSP-4, MGC9599, SP001LA, hBSSP-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62870