Learn More
Aviva Systems Biology Corp PRSS23 Antibody - C-terminal region (ARP52290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRSS23. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PRSS23. Alternate names: MGC5107, SIG13, SPUVE, ZSIG13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-687