Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10142MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Mammal. Species Reactivity: Mouse. Protein Expression: A DNA sequence encoding the Mus musculus (Mouse) Serine protease 29, was expressed in the hosts and tags indicated. AA Sequence: GTPAPGPEDV LMGIVGGHSA PQGKWPWQVS LRIYRYYWAF WVHNCGGSII HPQWVLTAAH CIRERDADPS VFRIRVGEAY LYGGKELLSV SRVIIHPDFV HAGLGSDVAL LQLAVSVQSF PNVKPVKLPS ESLEVTKKDV CWVTGWGAVS THRSLPPPYR LQQVQVKIID NSLCEEMYHN ATRHRNRGQK LILKDMLCAG NQGQDSCYGD SGGPLVCNVT GSWTLVGVVS WGYGCALRDF PGVYARVQSF LPWITQQMQR FS. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: Prss29. Gene ID: 114662. Uniprot: Q99MS4.
Catalog No. 50-188-5503