Learn More
Medchemexpress LLC PRSS3/Trypsin-3 Hum 10ug
SDP
Catalog No. 5000195719
Encompass_Preferred
Recombinant human PRSS3 (Trypsin-3) is a serine protease that cleaves peptide bonds after arginine residues. This product is supplied as 10 μg of recombinant, polyhistidine-tagged protein intended for enzymatic assays, inhibitor screening, and biochemical characterization.
  • Recombinant human enzyme for biochemical and enzymatic assays.
  • Polyhistidine tag facilitates purification and detection.
  • Supplied in a 10 μg unit size suitable for screening and pilot studies.
  • Appropriate for activity assays and inhibitor profiling.
  • Can be aliquoted and stored long term according to manufacturer instructions.
Catalog No. 50-001-95719 Supplier Medchemexpress LLC Supplier No. HYP7145610UG
Recombinant human PRSS3 (Trypsin-3) is a serine protease that cleaves peptide bonds after arginine residues. This product is supplied as 10 μg of recombinant, polyhistidine-tagged protein intended for enzymatic assays, inhibitor screening, and biochemical characterization.
  • Recombinant human enzyme for biochemical and enzymatic assays.
  • Polyhistidine tag facilitates purification and detection.
  • Supplied in a 10 μg unit size suitable for screening and pilot studies.
  • Appropriate for activity assays and inhibitor profiling.
  • Can be aliquoted and stored long term according to manufacturer instructions.