Learn More
Polysciences, Inc. Prussian Blue Iron Stain Kit (Reaction for Demonstration of Iron)
SDP
Catalog No. NC9934355
Encompass
Prussian Blue or Perls reaction is used to demonstrate ferric iron and ferritin. This is not a true staining technique rather, it is a histochemical reaction. The protein is split off by the hydrochloric acid, allowing the potassium ferrocyanide to combine with the ferric iron. This forms the ferric ferrocyanide or Prussian Blue.
Catalog No. NC9934355 Supplier Polysciences, Inc. Supplier No. 241991
Prussian Blue or Perls reaction is used to demonstrate ferric iron and ferritin. This is not a true staining technique rather, it is a histochemical reaction. The protein is split off by the hydrochloric acid, allowing the potassium ferrocyanide to combine with the ferric iron. This forms the ferric ferrocyanide or Prussian Blue.