Learn More
Medchemexpress LLC 2-[[(1R,2S)-2-aminocyclohexyl]amino]-4-[(3-methylphenyl)amino]-5-pyrimidinecarboxamide | 1194961-19-7 | 99.6% | 340.42 g/mol | C18H24N6O | 5 MG
SDP
Catalog No. 502030170
Encompass_Preferred
PRT-060318 (PRT318) is a potent, selective, and orally active inhibitor of spleen tyrosine kinase (Syk) intended for laboratory research. It exhibits low-nanomolar inhibitory potency and is provided as a high-purity solid with supporting analytical documentation for in vitro and in vivo studies.
  • Potent Syk inhibition with IC50 ≈ 3-4 nM.
  • High purity by HPLC (>99.5%).
  • Molecular formula C18H24N6O; molecular weight 340.42 g/mol.
  • Supplied as a 5 mg solid suitable for small-scale studies.
  • Analytical documentation available: datasheet, COA, SDS, NMR, LCMS.
  • For research use only; not for human or clinical use.
Catalog No. 50-203-0170 Supplier Medchemexpress LLC Supplier No. HY129745MG
PRT-060318 (PRT318) is a potent, selective, and orally active inhibitor of spleen tyrosine kinase (Syk) intended for laboratory research. It exhibits low-nanomolar inhibitory potency and is provided as a high-purity solid with supporting analytical documentation for in vitro and in vivo studies.
  • Potent Syk inhibition with IC50 ≈ 3-4 nM.
  • High purity by HPLC (>99.5%).
  • Molecular formula C18H24N6O; molecular weight 340.42 g/mol.
  • Supplied as a 5 mg solid suitable for small-scale studies.
  • Analytical documentation available: datasheet, COA, SDS, NMR, LCMS.
  • For research use only; not for human or clinical use.