Learn More
Medchemexpress LLC PRT062607 (acetate) | 1370261-98-5 | C21H27N9O3 | 1 ML
SDP
Catalog No. 5000372500
Encompass_Preferred
PRT062607 (acetate) is a laboratory chemical identified by CAS number 1370261-98-5 and chemical formula C21H27N9O3. It is stable under recommended storage conditions and intended for research use.
  • Solid appearance
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
  • Can be shipped at room temperature if less than 2 weeks
  • Also known as P505-15 (acetate), PRT-2607 (acetate), and BIIB-057 (acetate)
Catalog No. 50-003-72500 Supplier Medchemexpress LLC Supplier No. HY1532410MM/1ML
PRT062607 (acetate) is a laboratory chemical identified by CAS number 1370261-98-5 and chemical formula C21H27N9O3. It is stable under recommended storage conditions and intended for research use.
  • Solid appearance
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
  • Can be shipped at room temperature if less than 2 weeks
  • Also known as P505-15 (acetate), PRT-2607 (acetate), and BIIB-057 (acetate)