Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PRT062607 acetate | 1370261-98-5 | C21H27N9O3 | 100 MG
SDP

Catalog No. 5000372552
Encompass_Preferred

PRT062607 acetate is a solid chemical identified for laboratory use and substance manufacturing. It is recommended to store this product at 4°C, sealed and away from moisture. When in solvent, store at -80°C for up to 6 months or -20°C for 1 month, also sealed and away from moisture. Shipping at room temperature is acceptable for periods less than 2 weeks.

  • Identified for laboratory chemicals and manufacture of substances
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
  • Appearance: Solid

Catalog No. 50-003-72552 Supplier Medchemexpress LLC Supplier No. HY15324100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRT062607 acetate is a solid chemical identified for laboratory use and substance manufacturing. It is recommended to store this product at 4°C, sealed and away from moisture. When in solvent, store at -80°C for up to 6 months or -20°C for 1 month, also sealed and away from moisture. Shipping at room temperature is acceptable for periods less than 2 weeks.

  • Identified for laboratory chemicals and manufacture of substances
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
  • Appearance: Solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.