Learn More
Medchemexpress LLC HY-15323 5mg Medchemexpress, PRT062607 (Hydrochloride) CAS:1370261-97-4 Purity:>98%
SDP
Catalog No. 501932733
Encompass_Preferred
Medchemexpress, HY-15323 5mg PRT062607 (Hydrochloride) CAS:1370261-97-4 PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk ( IC 50 1-2 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2733 Supplier Medchemexpress LLC Supplier No. HY153235MG
Medchemexpress, HY-15323 5mg PRT062607 (Hydrochloride) CAS:1370261-97-4 PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk ( IC 50 1-2 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.