Learn More
Selleck Chemical LLC PRT4165 25mg 31083-55-3
SDP
Supplier:  Selleck Chemical LLC S531525MG
Encompass_Preferred
PRT4165 is a Bmi1/Ring1A inhibitor with an IC50 of 3.9 ¿M in the cell-free assay and also inhibits PRC1-mediated H2A ubiquitylation in vivo and in vitro. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8271