Learn More
TARGETMOL CHEMICALS INC PRT4165 50MG
SDP
Catalog No. 502576998
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PRT4165 (NSC600157) is a potent PRC1-mediated H2A ubiquitylation inhibitor. purity: 99%
Catalog No. 50-257-6998 Supplier TARGETMOL CHEMICALS INC Supplier No. T311050MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PRT4165 (NSC600157) is a potent PRC1-mediated H2A ubiquitylation inhibitor. purity: 99%