Learn More
MSC Heatstar Propane Convection Heater, 75,000 to 200,000 BTU, 4,700 sq. ft. Capacity, 1 ea.
Catalog No. 5000507803
Encompass
This 200,000 BTU convection propane industrial heater is designed for efficient, large-scale space heating. It utilizes natural convection to effectively warm areas up to 4,700 sq. ft. This unit is fully assembled and ready for immediate use, requiring no external electricity to operate.
  • Output range of 75,000 to 200,000 BTU per hour
  • Equipped with a thermoelectric safety valve for automatic gas shut-off
  • Factory-installed Piezo igniter for convenient lighting
  • Includes a 10 ft. propane hose and regulator assembly
  • Requires 100 lb. minimum propane tank size
  • Features standing pilot for increased safety
  • Clean burning operation
Catalog No. 50-005-07803 Supplier MSC Supplier No. 93178713
This 200,000 BTU convection propane industrial heater is designed for efficient, large-scale space heating. It utilizes natural convection to effectively warm areas up to 4,700 sq. ft. This unit is fully assembled and ready for immediate use, requiring no external electricity to operate.
  • Output range of 75,000 to 200,000 BTU per hour
  • Equipped with a thermoelectric safety valve for automatic gas shut-off
  • Factory-installed Piezo igniter for convenient lighting
  • Includes a 10 ft. propane hose and regulator assembly
  • Requires 100 lb. minimum propane tank size
  • Features standing pilot for increased safety
  • Clean burning operation