Learn More
MSC Goodway Portable Dry Steam Cleaner, 1700 W, 0.65 Gallon Boiler, 0-56 psi
Catalog No. 502836132
Encompass
This light-duty dry steam cleaner is designed for effective and eco-friendly cleaning in various commercial and institutional settings. This compact vapor steam cleaning and sanitizing system utilizes dry steam to eliminate grease and bacteria without chemicals. It is user-friendly, requiring only a plug-in to operate, with a lighting system indicating power status or refill needs.
  • Easy-to-use plug-and-go operation
  • Chemical-free cleaning eliminates grease and bacteria
  • 1700-watt boiler generates 311°F dry steam at the boiler
  • Removes oils, grease, and bacteria on contact
  • Includes an accessory kit for various surfaces and floors, and a long hose
  • Indicative lighting system shows power status or refill needs
  • Suitable for cleaning hotel rooms and bathrooms, eradicating bed bugs and pests, sanitizing small equipment, and maintaining production surfaces
Catalog No. 50-283-6132 Supplier MSC Supplier No. 59517318
This light-duty dry steam cleaner is designed for effective and eco-friendly cleaning in various commercial and institutional settings. This compact vapor steam cleaning and sanitizing system utilizes dry steam to eliminate grease and bacteria without chemicals. It is user-friendly, requiring only a plug-in to operate, with a lighting system indicating power status or refill needs.
  • Easy-to-use plug-and-go operation
  • Chemical-free cleaning eliminates grease and bacteria
  • 1700-watt boiler generates 311°F dry steam at the boiler
  • Removes oils, grease, and bacteria on contact
  • Includes an accessory kit for various surfaces and floors, and a long hose
  • Indicative lighting system shows power status or refill needs
  • Suitable for cleaning hotel rooms and bathrooms, eradicating bed bugs and pests, sanitizing small equipment, and maintaining production surfaces