Learn More
Rockland Immunochemicals Protein Molecular Weight Marker, 2000 uL
SDP
Catalog No. NC1758273
Encompass_Preferred
This protein molecular weight marker is a cocktail containing seven purified proteins that range in size from 14 to 120 kDa and thus span a molecular weight range common to most proteins and protein subunits. The protein mixture has been formulated to yield well-defined bands after SDS–polyacrylamide gel electrophoresis and staining with Coomassie blue or other common staining techniques.
Catalog No. NC1758273 Supplier Rockland Immunochemicals Supplier No. MB2012000
This protein molecular weight marker is a cocktail containing seven purified proteins that range in size from 14 to 120 kDa and thus span a molecular weight range common to most proteins and protein subunits. The protein mixture has been formulated to yield well-defined bands after SDS–polyacrylamide gel electrophoresis and staining with Coomassie blue or other common staining techniques.