Learn More
Aviva Systems Biology Corp PRTFDC1 Antibody - middle region (ARP48854_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48854_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRTFDC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRTFDC1. Alternate names: FLJ11888, HHGPApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59942