Learn More
Sino Biological Recombinant Human Proteinase 3 Protein (203S/A, His Tag), HPLC-verified
Supplier:  Sino Biological 13699H08H1100
Encompass_Preferred
A DNA sequence encoding the human PRTN3 (NP_002768.3, with mutation Ser 203 Ala) (Ile28-Pro256) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-211-5743