Learn More
Viva Products NoEndoTM Endotoxin Removal Spin Columns, Micro Capacity
SDP
Catalog No. NC2401952
Encompass
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 0.6 ml capacity; Micro Capacity resin can remove over 300 EU of endotoxin; 24/PK
Catalog No. NC2401952 Supplier Viva Products Supplier No. GENNOE24MICRO
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 0.6 ml capacity; Micro Capacity resin can remove over 300 EU of endotoxin; 24/PK