Learn More
Apexbio Technology LLC Prucalopride 179474-81-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062287
Encompass_Preferred
Prucalopride (CAS 179474-81-8) is a selective serotonin 5-HT4 receptor agonist with high affinity for both 5-HT4A and 5-HT4B receptor subtypes demonstrating Ki values of 2 5 nM and 8 nM respectively It displays substantial selectivity exceeding 290-fold against other serotonin receptor classes By activating 5-HT4 receptors prucalopride promotes gastrointestinal motility making it a valuable tool compound for investigating serotonergic mechanisms underlying gastrointestinal disorders and motility regulation Currently prucalopride is evaluated in Phase 3 clinical trials
Catalog No. 50-000-62287 Supplier Apexbio Technology LLC Supplier No. B22535.1
Prucalopride (CAS 179474-81-8) is a selective serotonin 5-HT4 receptor agonist with high affinity for both 5-HT4A and 5-HT4B receptor subtypes demonstrating Ki values of 2 5 nM and 8 nM respectively It displays substantial selectivity exceeding 290-fold against other serotonin receptor classes By activating 5-HT4 receptors prucalopride promotes gastrointestinal motility making it a valuable tool compound for investigating serotonergic mechanisms underlying gastrointestinal disorders and motility regulation Currently prucalopride is evaluated in Phase 3 clinical trials