Learn More
Medchemexpress LLC Prucalopride succinate | 179474-85-2 | 99.9% | 100 MG
SDP
Catalog No. 5000340898
Encompass_Preferred
Prucalopride succinate is an orally active, selective, and specific 5-HT4 receptor agonist with high affinity for human 5-HT4a/4b receptors. It improves intestinal motility by promoting regeneration of the intestinal nervous system in rats and shows anticancer activity by blocking the PI3K/AKT/mTor signaling pathway. This compound is suitable for research on chronic constipation, pseudo-intestinal obstruction, and cancer.
  • Orally active, selective, and specific 5-HT4 receptor agonist.
  • High affinity for human 5-HT4a/4b receptors (pKi values of 8.6 and 8.1, respectively).
  • Improves intestinal motility by promoting regeneration of the intestinal nervous system in rats.
  • Shows anticancer activity by blocking the PI3K/AKT/mTor signaling pathway.
  • Can be used in studies of chronic constipation, pseudo-intestinal obstruction, and cancer.
  • Available in solid form, white to off-white in color.
  • Purity of 99.9%.
Catalog No. 50-003-40898 Supplier Medchemexpress LLC Supplier No. HY12694100MG
Prucalopride succinate is an orally active, selective, and specific 5-HT4 receptor agonist with high affinity for human 5-HT4a/4b receptors. It improves intestinal motility by promoting regeneration of the intestinal nervous system in rats and shows anticancer activity by blocking the PI3K/AKT/mTor signaling pathway. This compound is suitable for research on chronic constipation, pseudo-intestinal obstruction, and cancer.
  • Orally active, selective, and specific 5-HT4 receptor agonist.
  • High affinity for human 5-HT4a/4b receptors (pKi values of 8.6 and 8.1, respectively).
  • Improves intestinal motility by promoting regeneration of the intestinal nervous system in rats.
  • Shows anticancer activity by blocking the PI3K/AKT/mTor signaling pathway.
  • Can be used in studies of chronic constipation, pseudo-intestinal obstruction, and cancer.
  • Available in solid form, white to off-white in color.
  • Purity of 99.9%.