Learn More
Medchemexpress LLC Prucalopride succinate | 179474-85-2 | 99.9% | 200 MG
SDP
Catalog No. 5000338716
Encompass_Preferred
Prucalopride succinate is an orally active, selective, and specific 5-HT4 receptor agonist. It enhances intestinal motility by promoting intestinal nervous system regeneration and exhibits anticancer activity by blocking the PI3K/AKT/mTor pathway. It is used in studies of chronic constipation, pseudo-intestinal obstruction, and cancer.
  • Selective and specific 5-HT4 receptor agonist
  • Enhances intestinal motility
  • Exhibits anticancer activity
  • Induces autophagy and apoptosis in A549/A427 cells
  • Promotes regeneration of colonic neural stem cells and neurons
Catalog No. 50-003-38716 Supplier Medchemexpress LLC Supplier No. HY12694200MG
Prucalopride succinate is an orally active, selective, and specific 5-HT4 receptor agonist. It enhances intestinal motility by promoting intestinal nervous system regeneration and exhibits anticancer activity by blocking the PI3K/AKT/mTor pathway. It is used in studies of chronic constipation, pseudo-intestinal obstruction, and cancer.
  • Selective and specific 5-HT4 receptor agonist
  • Enhances intestinal motility
  • Exhibits anticancer activity
  • Induces autophagy and apoptosis in A549/A427 cells
  • Promotes regeneration of colonic neural stem cells and neurons