Learn More
Selleck Chemical LLC Prucalopride Succinate S4247-50mg
SDP
Catalog No. 503111201
Encompass_Preferred
Prucalopride(R-108512) is a selective high affinity 5-HT4 receptor agonist The Ki values of prucalopride for human 5-HT(4a) and 5-HT(4b) receptor are 2 5 nM and 8 nM respectively
Catalog No. 50-311-1201 Supplier Selleck Chemical LLC Supplier No. S424750MG
Prucalopride(R-108512) is a selective high affinity 5-HT4 receptor agonist The Ki values of prucalopride for human 5-HT(4a) and 5-HT(4b) receptor are 2 5 nM and 8 nM respectively