Learn More
Medchemexpress LLC Prudomestin | 3443-28-5 | 330.29 | 5 MG
SDP
Catalog No. 5000415101
Encompass_Preferred
Prudomestin, isolated from the heartwood of *Prunus domestica*, exhibits potent xanthine oxidase (XO) inhibitory activity. It is suitable for research in inflammation and immunology.
  • Potent xanthine oxidase inhibitory activity with an IC50 of approximately 6 μM.
  • Isolated from the heartwood of *Prunus domestica*.
  • Has a molecular formula of C17H14O7.
  • Appears as a solid, light yellow to yellow.
  • Soluble in DMSO at ≥ 50 mg/mL.
  • Used in inflammation/immunology research, particularly in the study of metabolic enzymes.
Catalog No. 50-004-15101 Supplier Medchemexpress LLC Supplier No. HYN15475MG
Prudomestin, isolated from the heartwood of *Prunus domestica*, exhibits potent xanthine oxidase (XO) inhibitory activity. It is suitable for research in inflammation and immunology.
  • Potent xanthine oxidase inhibitory activity with an IC50 of approximately 6 μM.
  • Isolated from the heartwood of *Prunus domestica*.
  • Has a molecular formula of C17H14O7.
  • Appears as a solid, light yellow to yellow.
  • Soluble in DMSO at ≥ 50 mg/mL.
  • Used in inflammation/immunology research, particularly in the study of metabolic enzymes.