Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prudomestin | 3443-28-5 | 98.4% | 330.29 | C17H14O7 | 10 MG
SDP

Catalog No. 5000296532
Encompass_Preferred

Prudomestin is a flavonoid isolated from the heartwood of Prunus domestica that exhibits potent xanthine oxidase (XO) inhibitory activity (IC50 ≈ 6 μM). It is supplied as a light yellow to yellow solid for research use and should be protected from light during storage.

  • Potent xanthine oxidase inhibitor (IC50 ≈ 6 μM).
  • Molecular formula C17H14O7.
  • Molecular weight 330.29 g/mol.
  • CAS number 3443-28-5.
  • Purity 98.4%.
  • Appearance light yellow to yellow solid.
  • Storage recommendations: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-96532 Supplier Medchemexpress LLC Supplier No. HYN154710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prudomestin is a flavonoid isolated from the heartwood of Prunus domestica that exhibits potent xanthine oxidase (XO) inhibitory activity (IC50 ≈ 6 μM). It is supplied as a light yellow to yellow solid for research use and should be protected from light during storage.

  • Potent xanthine oxidase inhibitor (IC50 ≈ 6 μM).
  • Molecular formula C17H14O7.
  • Molecular weight 330.29 g/mol.
  • CAS number 3443-28-5.
  • Purity 98.4%.
  • Appearance light yellow to yellow solid.
  • Storage recommendations: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.