Learn More
Medchemexpress LLC HY-B0024 100mg Medchemexpress, Prulifloxacin CAS:123447-62-1 Purity:>98%
SDP
Catalog No. 502082347
Encompass_Preferred
Medchemexpress, HY-B0024 100mg Prulifloxacin CAS:123447-62-1 Prulifloxacin (NM441) is an orally active fluoroquinolone antibiotic with a broad spectrum of activity against Gram-positive and -negative bacteria. Prulifloxacin is a prodrug of a thiazeto-quinoline carboxylic acid derivative Ulifloxacin (NM394). Prulifloxacin has the potential for lower urinary tract infections and exacerbations of chronic bronchitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2347 Supplier Medchemexpress LLC Supplier No. HYB0024100MG
Medchemexpress, HY-B0024 100mg Prulifloxacin CAS:123447-62-1 Prulifloxacin (NM441) is an orally active fluoroquinolone antibiotic with a broad spectrum of activity against Gram-positive and -negative bacteria. Prulifloxacin is a prodrug of a thiazeto-quinoline carboxylic acid derivative Ulifloxacin (NM394). Prulifloxacin has the potential for lower urinary tract infections and exacerbations of chronic bronchitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.