Learn More
Cayman Chemical Prulifloxacin, Purity: ≥98%, CAS: 123447-62-1, 5 g
SDP
Catalog No. 502991040
Encompass_Preferred
A prodrug from of ulifloxacin; increases survival in various mouse models of systemic bacterial infection (ED50s = 0.35-23 mg/kg); increases survival in a mouse model of K. pneumoniae-induced respiratory tract infection (ED50 = 0.981 mg/kg)
Catalog No. 50-299-1040 Supplier Cayman Chemical Supplier No. 316245 G
A prodrug from of ulifloxacin; increases survival in various mouse models of systemic bacterial infection (ED50s = 0.35-23 mg/kg); increases survival in a mouse model of K. pneumoniae-induced respiratory tract infection (ED50 = 0.981 mg/kg)