Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prulifloxacin 98.0+%, TCI America™
SDP

Catalog No. P20581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20581G 1 g
P20585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20581G Supplier TCI America Supplier No. P20581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123447-62-1
Molecular Formula C21H20FN3O6S
Molecular Weight (g/mol) 461.46
MDL Number MFCD00864847
InChI Key PWNMXPDKBYZCOO-UHFFFAOYNA-N
PubChem CID 65947
IUPAC Name 6-fluoro-1-methyl-7-{4-[(5-methyl-2-oxo-2H-1,3-dioxol-4-yl)methyl]piperazin-1-yl}-4-oxo-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
SMILES CC1SC2=C(C(O)=O)C(=O)C3=CC(F)=C(C=C3N12)N1CCN(CC2=C(C)OC(=O)O2)CC1

Specifications

Melting Point 220°C
Color Yellow
Quantity 1 g
Formula Weight 461.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Prulifloxacin
RTECSNumber XJ0600000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.