Learn More
Medchemexpress LLC Prunasin | 99-18-3 | 295.29 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN15485MG
Encompass_Preferred
Prunasin is an orally active cyanogenic glucoside and the main metabolite of Amygdalin. It can specifically inhibit rat DNA polymerase β and exhibits anti-inflammatory and anti-fibrotic activities. Prunasin is used in the research of diseases such as liver fibrosis.
  • Orally active cyanogenic glucoside
  • Main metabolite of Amygdalin
  • Specifically inhibits rat DNA polymerase β (IC50: 98 μM)
  • Possesses anti-inflammatory activities
  • Possesses anti-fibrotic activities
  • Potentially useful in research for diseases like liver fibrosis
Catalog No. 50-004-15906