Learn More
Cusabio Technology LLC prune homolog Drosophila
Catalog No. 5000051472
Encompass_Preferred
This kit detects PRUNE (Protein prune homolog) in human serum plasma tissue homogenates and cell lysates using a quantitative sandwich ELISA method Designed for research applications it offers a detection range of 47 pg/mL to 3000 pg/mL with a sensitivity of 11 75 pg/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm ensuring reliable detection through standardized optical analysis Suitable for studies involving protein regulation cellular signaling or disease biomarker research this kit provides a robust tool for researchers requiring accurate quantification of PRUNE levels across diverse biological matrices
Catalog No. 50-000-51472 Supplier Cusabio Technology LLC Supplier No. CSBEL018831HU
This kit detects PRUNE (Protein prune homolog) in human serum plasma tissue homogenates and cell lysates using a quantitative sandwich ELISA method Designed for research applications it offers a detection range of 47 pg/mL to 3000 pg/mL with a sensitivity of 11 75 pg/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm ensuring reliable detection through standardized optical analysis Suitable for studies involving protein regulation cellular signaling or disease biomarker research this kit provides a robust tool for researchers requiring accurate quantification of PRUNE levels across diverse biological matrices