Learn More
Medchemexpress LLC HY-N1549 5mg Medchemexpress, Prunin CAS:529-55-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN15495MG
Encompass_Preferred
Medchemexpress, HY-N1549 5mg Prunin CAS:529-55-5 Prunin is a potent inhibitor of human enterovirus A71 (HEVA71). Prunin shows strong inhibitory activity against protein tyrosine phosphatase 1B (PTP1B), with an IC50 of 5.5 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2850