Learn More
MSC Milwaukee Tool Saw Chain and Guide Bar Set, 8 in, 3/8 in Low Profile Pitch, .043 in Gauge
Catalog No. 502837595
Encompass
This Saw Chain and Guide Bar Set provides an 8 in 3/8 in Low Profile Pitch, .043 in Gauge Saw Chain with a full house design for smooth pruning and clean finishes. It offers controlled, low-vibration cuts. The guide bar features a smaller radius nose for easy access in tight spaces and is engineered with lightweight materials for easy maneuvering. The narrow kerf, low profile cutters and guide bar are designed to provide more cuts per charge compared to standard kerf, maximizing battery efficiency.
  • Full house chain for smooth pruning and clean finishes.
  • Controlled, low-vibration cuts.
  • Smaller radius nose on guide bar for easy access in tight spaces.
  • Lightweight guide bar for easy maneuvering.
  • Narrow kerf, low profile cutters and guide bar provide more cuts per charge.
  • Low kickback saw chain.
  • Standard chain sequence.
Catalog No. 50-283-7595 Supplier MSC Supplier No. 51887461
This Saw Chain and Guide Bar Set provides an 8 in 3/8 in Low Profile Pitch, .043 in Gauge Saw Chain with a full house design for smooth pruning and clean finishes. It offers controlled, low-vibration cuts. The guide bar features a smaller radius nose for easy access in tight spaces and is engineered with lightweight materials for easy maneuvering. The narrow kerf, low profile cutters and guide bar are designed to provide more cuts per charge compared to standard kerf, maximizing battery efficiency.
  • Full house chain for smooth pruning and clean finishes.
  • Controlled, low-vibration cuts.
  • Smaller radius nose on guide bar for easy access in tight spaces.
  • Lightweight guide bar for easy maneuvering.
  • Narrow kerf, low profile cutters and guide bar provide more cuts per charge.
  • Low kickback saw chain.
  • Standard chain sequence.
Product Title