Learn More
MATTERHACKERS INC PRUSA XL 3D PRINTER FULLY ASSE
SDP
Catalog No. NC3705883
Encompass
  • Delivers consistent, high-resolution prints with exceptional reliability, even for intricate designs.
  • Optional tool changer allows integration of up to five materials or colors in a single print.
  • Large build volume of 360 mm x 360 mm x 360 mm accommodates extensive projects.
  • Nextruder's load cell sensor ensures automated first layer calibration for better print success.
  • Segmented heatbed enables localized heating, reducing energy consumption for smaller jobs.
Catalog No. NC3705883 Supplier MATTERHACKERS INC Supplier No. MW1S450L
  • Delivers consistent, high-resolution prints with exceptional reliability, even for intricate designs.
  • Optional tool changer allows integration of up to five materials or colors in a single print.
  • Large build volume of 360 mm x 360 mm x 360 mm accommodates extensive projects.
  • Nextruder's load cell sensor ensures automated first layer calibration for better print success.
  • Segmented heatbed enables localized heating, reducing energy consumption for smaller jobs.