Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prussian blue insoluble (iron(III) ferrocyanide) | 14038-43-8 | 99.0% | 859.23 | C18Fe7N18 | 250 G
SDP

Supplier:  Medchemexpress LLC HY106594A250G

Encompass_Preferred

Prussian blue insoluble (iron(III) ferrocyanide) is an inorganic pigment supplied as a fine, insoluble powder for laboratory and analytical use. It is commonly used as a reagent, pigment, and adsorbent where a stable, insoluble iron(III) ferrocyanide is required; batch-specific certificates of analysis are available.

  • 99.0% purity.
  • Molecular weight 859.23.
  • Chemical formula C18Fe7N18.
  • CAS number 14038-43-8.
  • Insoluble form suitable for pigment and adsorption applications.
  • Available in laboratory pack sizes up to 250 G.
  • Certificate of analysis provided for each batch.

Catalog No. 50-002-41031


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.