Learn More
Medchemexpress LLC Prussian blue insoluble (iron(III) ferrocyanide) | 14038-43-8 | MFCD00135663 | 99.0% | 859.23 | C18Fe7N18 | 100 G
SDP
Catalog No. 5000240301
Encompass_Preferred
Prussian blue insoluble (iron(III) ferrocyanide) is an insoluble pigment and adsorbent used as an antidote for radiocesium and thallium poisoning. It is investigated for antibacterial, anticancer, and imaging/photothermal applications, and is supplied as a high-purity fine powder for laboratory use.
  • Insoluble iron(III) ferrocyanide powder
  • Used as an antidote for cesium and thallium poisoning
  • Shows strong adsorbent properties for metal ions
  • Investigated for antibacterial and anticancer activity
  • Potential photoacoustic and MRI contrast agent
  • High purity suitable for research applications
  • Available in multiple laboratory package sizes
Catalog No. 50-002-40301 Supplier Medchemexpress LLC Supplier No. HY106594A100G
Prussian blue insoluble (iron(III) ferrocyanide) is an insoluble pigment and adsorbent used as an antidote for radiocesium and thallium poisoning. It is investigated for antibacterial, anticancer, and imaging/photothermal applications, and is supplied as a high-purity fine powder for laboratory use.
  • Insoluble iron(III) ferrocyanide powder
  • Used as an antidote for cesium and thallium poisoning
  • Shows strong adsorbent properties for metal ions
  • Investigated for antibacterial and anticancer activity
  • Potential photoacoustic and MRI contrast agent
  • High purity suitable for research applications
  • Available in multiple laboratory package sizes